Butamben (butyl-4-aminobenzoate) 98%
Supplier: Ambeed
|
Warning
|
A204115-100G
A204115-10G
A204115-250MG
A204115-500G
A204115-5G
A204115-25G
77030-224EA
26
USD
77030-224
77030-226
77030-228
77030-230
77030-232
77086-342
Butamben (butyl-4-aminobenzoate) 98%
Butamben (butyl-4-aminobenzoate)
Precaución: for Research Use Only
Formula:
C₁₁H₁₅NO₂ |
MDL Number:
MFCD00017112 CAS Number: 94-25-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...