2-Bromo-3-nitro-4-picoline 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-4-methyl-3-nitropyridine

A203955-100G A203955-10G A203955-1G A203955-25G A203955-5G
76999-430EA 350 USD
76999-430 76999-432 76999-434 76999-436 76999-438
2-Bromo-3-nitro-4-picoline 98%
2-Bromo-3-nitro-4-picoline

Precaución: for Research Use Only

Formula: C₆H₅BrN₂O₂
MW: 217.02
MDL Number: MFCD04112527
CAS Number: 23056-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR