3-Morpholin-4-ylbenzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-(3-Carboxyphenyl)morpholine, 3-Morpholinobenzoic acid, 3-(Morpholin-4-yl)benzoic acid

A203872-10G A203872-1G A203872-250MG A203872-5G
77012-736EA 159.14 USD
77012-736 77012-738 77012-740 77012-742
3-Morpholin-4-ylbenzoic acid 97%
3-Morpholin-4-ylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₃
MW: 207.23 g/mol
MDL Number: MFCD06659078
CAS Number: 215309-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR