6-Iodo-4-methoxy-1H-indazole 95%

Supplier: Ambeed
Warning

A203680-100MG A203680-250MG
77012-728EA 70.02 USD
77012-728 77012-730
6-Iodo-4-methoxy-1H-indazole 95%
6-Iodo-4-methoxy-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇IN₂O
MW: 274.06 g/mol
MDL Number: MFCD07781989
CAS Number: 885523-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR