1-(Pyrazin-2-yl)ethanamine 98%

Supplier: Ambeed
Warning

A203607-100MG A203607-1G A203607-250MG A203607-5G
77030-202EA 38.2 USD
77030-202 77030-204 77030-206 77030-208
1-(Pyrazin-2-yl)ethanamine 98%
1-(Pyrazin-2-yl)ethanamine

Precaución: for Research Use Only

Formula: C₆H₉N₃
MW: 123.15 g/mol
MDL Number: MFCD11506006
CAS Number: 179323-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR