2-(2,4-Diaminophenoxy)ethanol dihydrochloride 97%

Supplier: Ambeed
Warning

A203418-100G A203418-1G A203418-25G A203418-5G
76986-514EA 77.45 USD
76986-514 76986-516 76986-518 76986-520
2-(2,4-Diaminophenoxy)ethanol dihydrochloride 97%
2-(2,4-Diaminophenoxy)ethanol dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₄Cl₂N₂O₂
MW: 241.11 g/mol
MDL Number: MFCD00035494
CAS Number: 66422-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR