7-Hydroxy-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine 98%

Supplier: Ambeed
Warning

Synonyms: 7-Hydroxy-5-methyl-1,3,4-triazaindolizine, 5-Methyl-[1,2,4]triazolo[1,5-a]pyrimidin-7-ol, 4-Hydroxy-6-methyl-1,3,3a,7-tetraazaindene

A203397-100G A203397-10G A203397-25G A203397-500G A203397-5G
76999-420EA 73 USD
76999-420 76999-422 76999-424 76999-426 77881-481
7-Hydroxy-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine 98%
7-Hydroxy-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₆N₄O
MDL Number: MFCD00005555
CAS Number: 2503-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR