(R)-Piperidin-3-amine 97%

Supplier: Ambeed
Danger

A203214-1G A203214-250MG A203214-5G A203214-25G
76999-414EA 50 USD
76999-414 76999-416 76999-418 77881-480
(R)-Piperidin-3-amine 97%
(R)-Piperidin-3-amine

Precaución: for Research Use Only

Formula: C₅H₁₂N₂
MDL Number: MFCD03093369
CAS Number: 127294-73-9
UN: 3259
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR