(R)-tert-Butyl (4-oxo-4-(3-(trifluoromethyl)-5,6-dihydro-[1,2,4]triazolo[4,3-a]pyrazin-7(8H)-yl)-1-(2,4,5-trifluorophenyl)butan-2-yl)carbamate 95%

Supplier: Ambeed
Warning

Synonyms: N-Boc Sitagliptin

A203092-1G A203092-100MG
77030-194EA 260 USD
77030-194 77872-017
(R)-tert-Butyl (4-oxo-4-(3-(trifluoromethyl)-5,6-dihydro-[1,2,4]triazolo[4,3-a]pyrazin-7(8H)-yl)-1-(2,4,5-trifluorophenyl)butan-2-yl)carbamate 95%
(R)-tert-Butyl (4-oxo-4-(3-(trifluoromethyl)-5,6-dihydro-[1,2,4]triazolo[4,3-a]pyrazin-7(8H)-yl)-1-(2,4,5-trifluorophenyl)butan-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₂₁H₂₃F₆N₅O₃
MDL Number: MFCD29044848
CAS Number: 486460-23-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR