Methyl 3-hydroxy-4,5-dimethylthiophene-2-carboxylate 98%

Supplier: Ambeed
Warning

A202969-1G A202969-250MG
77030-188EA 150.65 USD
77030-188 77030-190
Methyl 3-hydroxy-4,5-dimethylthiophene-2-carboxylate 98%
Methyl 3-hydroxy-4,5-dimethylthiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₀O₃S
MW: 186.23 g/mol
MDL Number: MFCD15071706
CAS Number: 32822-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR