3-(Hydroxymethyl)-1-methyl-1H-indazole 95%

Supplier: Ambeed
Warning

A202553-1G A202553-250MG
77030-170EA 75 USD
77030-170 77030-172
3-(Hydroxymethyl)-1-methyl-1H-indazole 95%
3-(Hydroxymethyl)-1-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O
MDL Number: MFCD08060470
CAS Number: 1578-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR