N-(4-Methoxybenzyl)propan-2-amine 95%

Supplier: Ambeed
Warning

A202496-250MG A202496-1G A202496-5G
77012-702EA 20 USD
77012-702 77890-225 77890-226
N-(4-Methoxybenzyl)propan-2-amine 95%
N-(4-Methoxybenzyl)propan-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO
MDL Number: MFCD06740633
CAS Number: 70894-74-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR