N,N-Diethyl 3-borono-4-methylbenzenesulfonamide 98%

Supplier: Ambeed
Warning

Synonyms: (5-(N,N-Diethylsulphamoyl)-2-methylphenyl)boronic acid, 5-Diethylsulphamoyl-2-methylbenzeneboronic acid

A202398-1G A202398-250MG
77012-698EA 165.51 USD
77012-698 77012-700
N,N-Diethyl 3-borono-4-methylbenzenesulfonamide 98%
N,N-Diethyl 3-borono-4-methylbenzenesulphonamide

Precaución: for Research Use Only

Formula: C₁₁H₁₈BNO₄S
MW: 271.14 g/mol
MDL Number: MFCD08056356
CAS Number: 871329-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR