Methyl 4-amino-5-methylthiophene-2-carboxylate 97%

Supplier: Ambeed
Warning

A202332-100MG A202332-250MG A202332-5G
77012-688EA 25.47 USD
77012-688 77012-690 77012-692
Methyl 4-amino-5-methylthiophene-2-carboxylate 97%
Methyl 4-amino-5-methylthiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉NO₂S
MW: 171.217 g/mol
MDL Number: MFCD14694722
CAS Number: 501082-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR