2-Chloro-5-nitro-3-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Danger

Synonyms: 2-Chloro-5-nitro-3-(trifluoromethyl)pyridine

A202273-1G A202273-250MG A202273-5G A202273-25G
76999-398EA 424.36 USD
76999-398 76999-400 76999-402 76999-404 77881-476
2-Chloro-5-nitro-3-(trifluoromethyl)pyridine 98%
2-Chloro-5-nitro-3-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₂ClF₃N₂O₂
MW: 226.54 g/mol
MDL Number: MFCD00792435
CAS Number: 99368-67-9
UN: 2922
ADR: 8,6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR