3-Phenoxypyrrolidine hydrochloride 97%

Supplier: Ambeed
Warning

A202269-100MG A202269-1G A202269-250MG
76999-394EA 72 USD
76999-394 76999-396 77671-796
3-Phenoxypyrrolidine hydrochloride 97%
3-Phenoxypyrrolidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₄ClNO
MDL Number: MFCD09879231
CAS Number: 21767-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR