Methyl-2-amino-4-bromobenzoate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Amino-4-bromobenzoic acid methyll ester, Methyl 2-amino-4-bromobenzoate
A202155-100G
A202155-10G
A202155-1G
A202155-25G
A202155-500G
A202155-5G
76999-382EA
157.02
USD
76999-382
76999-384
76999-386
76999-388
76999-390
76999-392
Methyl-2-amino-4-bromobenzoate 97%
Methyl 2-amino-4-bromobenzoate
Precaución: for Research Use Only
Formula:
C₈H₈BrNO₂ MW: 230.0586 g/mol |
MDL Number:
MFCD07783014 CAS Number: 135484-83-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...