2,7-Dibromo-9,9-didecylfluorene 97%

Supplier: Ambeed
Warning

A202147-10G A202147-25G A202147-5G
77030-164EA 21 USD
77030-164 77030-166 77030-168
2,7-Dibromo-9,9-didecylfluorene 97%
2,7-Dibromo-9,9-didecylfluorene

Precaución: for Research Use Only

Formula: C₃₃H₄₈Br₂
MDL Number: MFCD00971288
CAS Number: 175922-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR