(1S,3aR,6aS)-Ethyl octahydrocyclopenta[c]pyrrole-1-carboxylate hydrochloride 95%

Supplier: Ambeed
Warning

A202016-100G A202016-10G A202016-1G A202016-250MG A202016-25G A202016-5G
77012-666EA 1623.18 USD
77012-666 77012-668 77012-670 77012-672 77012-674 77012-676
(1S,3aR,6aS)-Ethyl octahydrocyclopenta[c]pyrrole-1-carboxylate hydrochloride 95%
(1S,3aR,6aS)-Ethyl octahydrocyclopenta[c]pyrrole-1-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₈ClNO₂
MW: 219.71 g/mol
MDL Number: MFCD21606967
CAS Number: 1147103-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR