3-Cyano-5-methylbenzoic acid 98%

Supplier: Ambeed
Warning

A201945-100MG A201945-1G A201945-250MG A201945-5G
76986-496EA 16 USD
76986-496 76986-498 76986-500 77885-471
3-Cyano-5-methylbenzoic acid 98%
3-Cyano-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₂
MDL Number: MFCD11007822
CAS Number: 78621-81-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR