1-(6-Chloro-5-methylpyridin-3-yl)ethan-1-one 97%

Supplier: Ambeed
Warning

Synonyms: 1-(6-Chloro-5-methylpyridin-3-yl)ethanone

A201802-100MG A201802-1G A201802-250MG
77012-660EA 120.95 USD
77012-660 77012-662 77012-664
1-(6-Chloro-5-methylpyridin-3-yl)ethan-1-one 97%
1-(6-Chloro-5-methylpyridin-3-yl)ethan-1-one

Precaución: for Research Use Only

Formula: C₈H₈ClNO
MW: 169.608 g/mol
MDL Number: MFCD18258044
CAS Number: 1256791-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR