2-Ethylhexyl-2-cyano-3,3-diphenylacrylate 98%

Supplier: Ambeed
Warning

Synonyms: 2-Ethylhexyl 2-cyano-3,3-diphenylacrylate, Octyl 2-Cyano-3,3-diphenylacrylate, 2-Cyano-3,3-diphenylacrylic Acid 2-Ethylhexyl Ester, Octocrylene

A201796-1000G A201796-100G A201796-25G A201796-500G A201796-5G
77030-154EA 154.9 USD
77030-154 77030-156 77030-158 77030-160 77030-162
2-Ethylhexyl-2-cyano-3,3-diphenylacrylate 98%
2-Ethylhexyl-2-cyano-3,3-diphenylacrylate

Precaución: for Research Use Only

Formula: C₂₄H₂₇NO₂
MW: 361.49 g/mol
MDL Number: MFCD00059260
CAS Number: 6197-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR