Ethyl 1H-pyrrolo[2,3-c]pyridine-2-carboxylate 95%

Supplier: Ambeed
Warning

A201769-100MG A201769-1G A201769-250MG A201769-5G
77030-146EA 39.26 USD
77030-146 77030-148 77030-150 77030-152
Ethyl 1H-pyrrolo[2,3-c]pyridine-2-carboxylate 95%
Ethyl 1H-pyrrolo[2,3-c]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MW: 190.199 g/mol
MDL Number: MFCD11506124
CAS Number: 24334-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR