AMI-1 98%

Supplier: Ambeed
Warning

Synonyms: AMI 1, Sodium 7,7'-(carbonylbis(azanediyl))bis(4-hydroxynaphthalene-2-sulphonate), 7,7'-(Carbonyldiimino)bis[4-hydroxy2-naphthalenesulphonicacid]disodiumsalt, 7,7′-Carbonylbis(azanediyl)bis(4-hydroxynaphthalene-2-sulfonic acid) sodium salt, AMI-1 sodium salt

A201673-10MG A201673-1MG A201673-25MG A201673-5MG
76999-338EA 21.99 USD
76999-338 76999-340 76999-342 76999-344
AMI-1 98%
AMI-1

Precaución: for Research Use Only

Formula: C₂₁H₁₄N₂Na₂O₉S₂
MDL Number: MFCD00068249
CAS Number: 20324-87-2 | 134-47-4 (free base)

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR