Methyl 2-bromo-4-(2-methoxy-2-oxoethyl)thiazole-5-carboxylate 95%

Supplier: Ambeed
Warning

A201625-100MG A201625-1G A201625-250MG
77012-654EA 21.22 USD
77012-654 77012-656 77012-658
Methyl 2-bromo-4-(2-methoxy-2-oxoethyl)thiazole-5-carboxylate 95%
Methyl 2-bromo-4-(2-methoxy-2-oxoethyl)thiazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₄S
MW: 294.12 g/mol
MDL Number: MFCD13195322
CAS Number: 365996-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR