7-Nitroindole-3-carboxyaldehyde 95%

Supplier: Ambeed
Warning

A201139-1G A201139-5G
76999-326EA 22 USD
76999-326 76999-328
7-Nitroindole-3-carboxyaldehyde 95%
7-Nitroindole-3-carboxyaldehyde

Precaución: for Research Use Only

Formula: C₉H₆N₂O₃
MDL Number: MFCD01074519
CAS Number: 10553-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR