Boc-D-ORN-OH 98%

Supplier: Ambeed
Warning

A201059-250MG A201059-5G A201059-1G
76986-470EA 15 USD
76986-470 76986-472 77593-302
Boc-D-ORN-OH 98%
Boc-D-ORN-OH

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₄
MDL Number: MFCD00236869
CAS Number: 159877-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR