5-Chloro-2-ethylpyridine 95%

Supplier: Ambeed
Warning

A200950-100MG A200950-1G A200950-250MG
76986-464EA 84.88 USD
76986-464 76986-466 76986-468
5-Chloro-2-ethylpyridine 95%
5-Chloro-2-ethylpyridine

Precaución: for Research Use Only

Formula: C₇H₈ClN
MW: 141.60 g/mol
MDL Number: MFCD12033380
CAS Number: 1159815-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR