N-Ethyl-4-bromobenzylamine 97%

Supplier: Ambeed
Warning

A200867-10G A200867-1G A200867-5G
77012-640EA 89.12 USD
77012-640 77012-642 77012-644
N-Ethyl-4-bromobenzylamine 97%
N-Ethyl-4-bromobenzylamine

Precaución: for Research Use Only

Formula: C₉H₁₂BrN
MW: 214.10 g/mol
MDL Number: MFCD07404997
CAS Number: 856795-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR