2-Cyclopropylpyridin-4-amine 98%

Supplier: Ambeed
Warning

A200866-100MG A200866-50MG
76986-460EA 105.03 USD
76986-460 76986-462
2-Cyclopropylpyridin-4-amine 98%
2-Cyclopropylpyridin-4-amine

Precaución: for Research Use Only

Formula: C₈H₁₀N₂
MW: 134.178 g/mol
MDL Number: MFCD12032771
CAS Number: 340006-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR