tert-Butyl-4-hydroxy-2-oxopiperidine-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-hydroxy-2-oxopiperidine-1-carboxylate

A200844-1G A200844-250MG A200844-5G
77030-122EA 221.73 USD
77030-122 77030-124 77030-126
tert-Butyl-4-hydroxy-2-oxopiperidine-1-carboxylate 97%
tert-Butyl-4-hydroxy-2-oxopiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₄
MW: 215.2500 g/mol
MDL Number: MFCD16659601
CAS Number: 1245646-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR