3,3-Dimethyl-5-nitroindolin-2-one 95%

Supplier: Ambeed
Warning

A200678-100MG A200678-250MG
76986-450EA 66.84 USD
76986-450 76986-452
3,3-Dimethyl-5-nitroindolin-2-one 95%
3,3-Dimethyl-5-nitroindolin-2-one

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₃
MW: 206.198 g/mol
MDL Number: MFCD13191795
CAS Number: 100511-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR