4-Methoxy-1-benzothiophene-2-carboxylic acid 96%

Supplier: Ambeed
Warning

A200613-100MG A200613-250MG A200613-1G A200613-5G
77012-630EA 17 USD
77012-630 77611-946 77890-215 77890-216
4-Methoxy-1-benzothiophene-2-carboxylic acid 96%
4-Methoxy-1-benzothiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₈O₃S
MDL Number: MFCD07371550
CAS Number: 476199-14-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR