2,7-Dibromo-9-(9-heptadecyl)carbazole 95%

Supplier: Ambeed
Warning

A200609-100MG A200609-250MG A200609-1G
76986-430EA 6 USD
76986-430 76986-432 77885-467
2,7-Dibromo-9-(9-heptadecyl)carbazole 95%
2,7-Dibromo-9-(9-heptadecyl)carbazole

Precaución: for Research Use Only

Formula: C₂₉H₄₁Br₂N
MDL Number: MFCD16619379
CAS Number: 955964-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR