6-Bromo-3H-thieno[2,3-d]pyrimidin-4-one 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Bromothieno[2,3-d]pyrimidin-4(3H)-one
A200470-1G
A200470-250MG
A200470-5G
77030-106EA
79.57
USD
77030-106
77030-108
77567-894
6-Bromo-3H-thieno[2,3-d]pyrimidin-4-one 95%
6-Bromo-3H-thieno[2,3-d]pyrimidin-4-one
Precaución: For research use only.
Formula:
C₆H₃BrN₂OS MW: 231.07 g/mol |
MDL Number:
MFCD12196499 CAS Number: 56844-40-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...