2-Chloro-4-methylphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-4-methylbenzeneboronic acid

A200438-25G A200438-1G
76999-310EA 80.63 USD
76999-310 77671-784
2-Chloro-4-methylphenylboronic acid 98%
2-Chloro-4-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BClO₂
MW: 170.40 g/mol
MDL Number: MFCD03411936
CAS Number: 145349-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR