(R)-1-Boc-3-(Dimethylamino)pyrrolidine 98%

Supplier: Ambeed
Warning

A200376-1G A200376-250MG A200376-5G
77030-100EA 74 USD
77030-100 77030-102 77030-104
(R)-1-Boc-3-(Dimethylamino)pyrrolidine 98%
(R)-1-Boc-3-(Dimethylamino)pyrrolidine

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MDL Number: MFCD11053517
CAS Number: 1004538-33-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR