4-(Phenylthio)benzaldehyde 95%

Supplier: Ambeed
Warning

A200374-10G A200374-1G A200374-5G A200374-25G A200374-100G
76999-304EA 88.06 USD
76999-304 76999-306 76999-308 77600-624 77671-776
4-(Phenylthio)benzaldehyde 95%
4-(Phenylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₀OS
MW: 214.28 g/mol
MDL Number: MFCD01666578
CAS Number: 1208-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR