(3-Amino-5-chlorophenyl)methanol 95%

Supplier: Ambeed
Warning

A200305-250MG A200305-1G
77030-098EA 43.5 USD
77030-098 77667-656
(3-Amino-5-chlorophenyl)methanol 95%
(3-Amino-5-chlorophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₈ClNO
MW: 157.6 g/mol
MDL Number: MFCD11215511
CAS Number: 79944-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR