6-Methyl-1-indanone 98%

Supplier: Ambeed
Warning

A200211-100G A200211-1G A200211-25G A200211-5G
76999-296EA 216.43 USD
76999-296 76999-298 76999-300 76999-302
6-Methyl-1-indanone 98%
6-Methyl-1-indanone

Precaución: for Research Use Only

Formula: C₁₀H₁₀O
MW: 146.19 g/mol
MDL Number: MFCD00275729
CAS Number: 24623-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR