1-Chloro-4-methylisoquinoline 97%

Supplier: Ambeed
Warning

A200123-100MG A200123-250MG A200123-1G
77012-620EA 45 USD
77012-620 77012-622 77611-934
1-Chloro-4-methylisoquinoline 97%
1-Chloro-4-methylisoquinoline

Precaución: for Research Use Only

Formula: C₁₀H₈ClN
MW: 177.63 g/mol
MDL Number: MFCD18448753
CAS Number: 24188-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR