2-Chloro-N-fmoc-L-phenylalanine 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Phe(2-Cl)-OH

A200110-1G A200110-250MG A200110-5G A200110-25G
76999-292EA 27 USD
76999-292 76999-294 77671-770 77881-466
2-Chloro-N-fmoc-L-phenylalanine 95%
2-Chloro-N-fmoc-L-phenylalanine

Precaución: for Research Use Only

Formula: C₂₄H₂₀ClNO₄
MW: 421.877 g/mol
MDL Number: MFCD00797578
CAS Number: 198560-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR