4-(2-Methoxyethyl)piperidine hydrochloride 98+%
Supplier: Ambeed
|
Warning
|
A200071-1G
A200071-250MG
76999-282EA
68.96
USD
76999-282
76999-284
4-(2-Methoxyethyl)piperidine hydrochloride 98+%
4-(2-Methoxyethyl)piperidine hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₈ClNO |
MDL Number:
MFCD10687230 CAS Number: 868849-54-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...