2-Anilinobenzoic acid methyl ester 98%

Supplier: Ambeed
Warning

A199957-1G A199957-250MG A199957-5G
77012-614EA 56.23 USD
77012-614 77012-616 77012-618
2-Anilinobenzoic acid methyl ester 98%
2-Anilinobenzoic acid methyl ester

Precaución: for Research Use Only

Formula: C₁₄H₁₃NO₂
MW: 227.26 g/mol
MDL Number: MFCD01366532
CAS Number: 35708-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR