(S)-(-)-2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole 98%

Supplier: Ambeed
Warning

Synonyms: (S)-N-Despropyl Pramipexole, Pramipexole Related Compound A

A199923-10G A199923-25G A199923-5G A199923-500G A199923-100G A199923-1G
76999-276EA 20.16 USD
76999-276 76999-278 76999-280 77671-768 77671-764 77671-766
(S)-(-)-2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole 98%
(S)-(-)-2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole

Precaución: for Research Use Only

Formula: C₇H₁₁N₃S
MW: 169.25 g/mol
MDL Number: MFCD07368003
CAS Number: 106092-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR