AS-703026 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-N-(2,3-Dihydroxypropyl)-3-((2-fluoro-4-iodophenyl)amino)isonicotinamide, MSC1936369B, Pimasertib
A199908-10MG
A199908-50MG
A199908-5MG
A199908-1MG
A199908-250MG
A199908-1G
77012-608EA
15.92
USD
77012-608
77012-610
77012-612
77611-924
77611-926
77683-555
AS-703026 98%
AS-703026
Precaución: For research use only.
Formula:
C₁₅H₁₅FIN₃O₃ MW: 431.2 g/mol |
MDL Number:
MFCD16660676 CAS Number: 1236699-92-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...