6-(Trifluoromethyl)imidazo[2,1-b]thiazole-5-carbaldehyde 98%

Supplier: Ambeed
Warning

A199690-100MG A199690-1G
76986-418EA 38.2 USD
76986-418 76986-420
6-(Trifluoromethyl)imidazo[2,1-b]thiazole-5-carbaldehyde 98%
6-(Trifluoromethyl)imidazo[2,1-b]thiazole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₇H₃F₃N₂OS
MW: 220.17 g/mol
MDL Number: MFCD05179682
CAS Number: 564443-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR