4-Bromo-2-(difluoromethyl)pyridine 95%

Supplier: Ambeed
Danger

A199625-100MG A199625-1G A199625-250MG A199625-5G A199625-25G
76986-410EA 7 USD
76986-410 76986-412 76986-414 76986-416 77885-464
4-Bromo-2-(difluoromethyl)pyridine 95%
4-Bromo-2-(difluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₄BrF₂N
MDL Number: MFCD18257229
CAS Number: 1211580-54-9
UN: 1759
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR