6-Aminotetrahydro-2,2-dimethyl-4H-cyclopenta-1,3-dioxol-4-ol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ticagrelor Related Compound 1, (3aR,4S,6R,6aS)-6-Amino-2,2-dimethyltetrahydro-3aH-cyclopenta[d][1,3]dioxol-4-ol, (3aR,4S,6R,6aS)-6-amino-2,2-dimethyl-hexahydrocyclopenta[d][1,3]dioxol-4-ol
A199601-100G
A199601-10G
A199601-1G
A199601-5G
A199601-25G
76986-402EA
217.49
USD
76986-402
76986-404
76986-406
76986-408
77611-920
6-Aminotetrahydro-2,2-dimethyl-4H-cyclopenta-1,3-dioxol-4-ol 97%
6-Aminotetrahydro-2,2-dimethyl-4H-cyclopenta-1,3-dioxol-4-ol
Precaución: for Research Use Only
Formula:
C₈H₁₅NO₃ MW: 173.21 g/mol |
MDL Number:
MFCD20527303 CAS Number: 155899-66-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...