3-Bromo-5,6,7,8-tetrahydroquinoline 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A199488-100MG A199488-1G A199488-250MG
76986-396EA 54.11 USD
76986-396 77683-553 77683-554
3-Bromo-5,6,7,8-tetrahydroquinoline 95%, Ambeed.Inc
3-Bromo-5,6,7,8-tetrahydroquinoline

Precaución: For research use only.

Formula: C₉H₁₀BrN
MW: 212.08 g/mol
MDL Number: MFCD11111668
CAS Number: 82132-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR